accessibility menu, dialog, popup

UserName

CH3OOHBOH3CH3CH3CCH3O

CAS RN 269410-23-3

IUPAC Name: 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 250.1
Molecular Formula: C13H19BO4
InChi Key: WINSAMQIAMEZIK-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1O)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results