accessibility menu, dialog, popup

UserName

IOO

CAS RN 2689-65-8

IUPAC Name: 5-iodofuran-2-carbaldehyde
Synonyms: 5-iodofuran-2-carbaldehyde
Molecular Weight (g/mol): 221.98
Molecular Formula: C5H3IO2
InChi Key: QPGPCPKDVSPJAY-UHFFFAOYSA-N
SMILES: IC1=CC=C(O1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results