accessibility menu, dialog, popup

UserName

SSNN

CAS RN 2645-22-9

IUPAC Name: 4-(pyridin-4-yldisulfanyl)pyridine
Synonyms: 4,4'-dipyridyl disulfide
Molecular Weight (g/mol): 220.31
Molecular Formula: C10H8N2S2
InChi Key: UHBAPGWWRFVTFS-UHFFFAOYSA-N
SMILES: S(SC1=CC=NC=C1)C1=CC=NC=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results