CAS RN 264272-46-0
CAS RN 264272-46-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
4-(1,1-Dimethylethyl)-a-[hydroxy(1,3,4-trimethyl-1H-pyrazol-5-yl)methylene]benzeneacetonitrile (E/Z mixture), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(1,1-Dimethylethyl)-Alpha-[hydroxy(1,3,4-trimethyl-1H-pyrazol-5-yl)methylene]benzeneacetonitrile (E/Z mixture), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(1,1-Dimethylethyl)-a-[hydroxy(1,3,4-trimethyl-1H-pyrazol-5-yl)methylene]benzeneacetonitrile (E/Z mixture), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.