accessibility menu, dialog, popup

UserName

CAS RN 25961-11-9

H3CONCl

CAS RN 25961-11-9

IUPAC Name: 1-{5-chloro-2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}ethan-1-one
Synonyms: 1-{5-chloro-2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}ethanone
Molecular Weight (g/mol): 271.74
Molecular Formula: C16H14ClNO
InChi Key: NMZOSOMVILZBJL-UHFFFAOYSA-N
SMILES: CC(=O)N1C2=CC=CC=C2CCC2=CC=C(Cl)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results