accessibility menu, dialog, popup

UserName

OOOHCl

CAS RN 2591-25-5

IUPAC Name: (6-chloro-2H-1,3-benzodioxol-5-yl)methanol
Synonyms: (6-chloro-2H-1,3-benzodioxol-5-yl)methanol
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: ADHYYNHCXPPVHQ-UHFFFAOYSA-N
SMILES: OCC1=CC2=C(OCO2)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results