accessibility menu, dialog, popup

UserName

CAS RN 258506-49-9

NOOBrCl

CAS RN 258506-49-9

IUPAC Name: 2-bromo-1-[3-(4-chlorophenyl)-1,2-oxazol-5-yl]ethan-1-one
Synonyms: 2-bromo-1-[3-(4-chlorophenyl)-1,2-oxazol-5-yl]ethanone
Molecular Weight (g/mol): 300.54
Molecular Formula: C11H7BrClNO2
InChi Key: OUHGJKFYGYXITB-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=NOC(=C1)C(=O)CBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results