accessibility menu, dialog, popup

UserName

CAS RN 25570-02-9

CH3H3CCH3CH3

CAS RN 25570-02-9

IUPAC Name: 3,3',5,5'-tetramethyl-1,1'-biphenyl
Synonyms: 3,3',5,5'-tetramethyl-1,1'-biphenyl
Molecular Weight (g/mol): 210.32
Molecular Formula: C16H18
InChi Key: CMZYGFLOKOQMKF-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)C1=CC(C)=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results