CAS RN 2555-28-4
CAS RN 2555-28-4
IUPAC Name:
7-methoxy-4-methyl-2H-chromen-2-one
Synonyms:
coumarin,4-methyl-7-methoxy
Molecular Weight (g/mol):
190.2
Molecular Formula:
C11H10O3
InChi Key:
UDFPKNSWSYBIHO-UHFFFAOYSA-N
SMILES:
COC1=CC=C2C(C)=CC(=O)OC2=C1
1
–
2
of
2
results
1
7-Methoxy-4-methylcoumarin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Methoxy-4-methylcoumarin, 99%, Thermo Scientific™
CAS: 2555-28-4 Molecular Formula: C11H10O3 Molecular Weight (g/mol): 190.198 MDL Number: MFCD00009773 InChI Key: UDFPKNSWSYBIHO-UHFFFAOYSA-N Synonym: 7-methoxy-4-methylcoumarin,7-methoxy-4-methyl-2h-chromen-2-one,2h-1-benzopyran-2-one, 7-methoxy-4-methyl,4-methylherniarin,hymecromone methyl ether,herniarin, 4-methyl,7-methoxy-4-methyl-chromen-2-one,4-methyl-7-methoxycoumarin,coumarin,4-methyl-7-methoxy,coumarin, 7-methoxy-4-methyl PubChem CID: 390807 IUPAC Name: 7-methoxy-4-methylchromen-2-one SMILES: CC1=CC(=O)OC2=C1C=CC(=C2)OC