CAS RN 25135-51-7
CAS RN 25135-51-7
IUPAC Name:
4-(4-hydroxybenzenesulfonyl)phenol; 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
Synonyms:
bisphenol S; bisphenol-A
Molecular Weight (g/mol):
478.56
Molecular Formula:
C27H26O6S
InChi Key:
ZUZZUJNBGCVPLQ-UHFFFAOYSA-N
SMILES:
CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1.OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
Polysulfone Resin, Pellets
CAS: 25135-51-7 Molecular Formula: (C27H22O4S)n Molecular Weight (g/mol): NaN MDL Number: MFCD00134387 InChI Key: ZUZZUJNBGCVPLQ-UHFFFAOYSA-N PubChem CID: 16211415 SMILES: CC(C)(C1=CC=C(O-*)C=C1)C1=CC=C(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(-*)C=C2)C=C1