accessibility menu, dialog, popup

UserName

OFFNH2

CAS RN 244022-71-7

IUPAC Name: 1-[3-(difluoromethoxy)phenyl]methanamine
Synonyms: 1-[3-(difluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol): 173.16
Molecular Formula: C8H9F2NO
InChi Key: SYAQBBDPVPDVLJ-UHFFFAOYSA-N
SMILES: NCC1=CC(OC(F)F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results