accessibility menu, dialog, popup

UserName

BrNO

CAS RN 243455-57-4

IUPAC Name: 5-(3-bromophenyl)-1,3-oxazole
Synonyms: 5-(3-bromophenyl)-1,3-oxazole
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: GUYZQMXSOVOSIC-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=CN=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results