CAS RN 241479-74-3
CAS RN 241479-74-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(AlphaS,BetaR)-Beta-(2,5-Difluorophenyl)-Beta-hydroxy-Alpha-methyl-1H-1,2,4-triazole-1-butanenitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(AlphaS,BetaR)-Beta-(2,5-Difluorophenyl)-Beta-hydroxy-Alpha-methyl-1H-1,2,4-triazole-1-butanenitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.