accessibility menu, dialog, popup

UserName

CAS RN 2404-35-5

IUPAC Name: bromocycloheptane
Synonyms: bromocycloheptane
Molecular Weight (g/mol): 177.09
Molecular Formula: C7H13Br
InChi Key: LOXORFRCPXUORP-UHFFFAOYSA-N
SMILES: BrC1CCCCCC1
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)