accessibility menu, dialog, popup

UserName

FFFSNCO

CAS RN 24032-84-6

IUPAC Name: 1-isocyanato-4-[(trifluoromethyl)sulfanyl]benzene
Synonyms: 1-isocyanato-4-[(trifluoromethyl)sulfanyl]benzene
Molecular Weight (g/mol): 219.18
Molecular Formula: C8H4F3NOS
InChi Key: NYQSIHZNEJCZKX-UHFFFAOYSA-N
SMILES: FC(F)(F)SC1=CC=C(C=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results