accessibility menu, dialog, popup

UserName

CH3OFFFF

CAS RN 239107-27-8

IUPAC Name: 1-[4-fluoro-3-(trifluoromethyl)phenyl]propan-1-one
Synonyms: 1-[4-fluoro-3-(trifluoromethyl)phenyl]propan-1-one
Molecular Weight (g/mol): 220.17
Molecular Formula: C10H8F4O
InChi Key: DEKWUKRLNDKROW-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC(=C(F)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results