accessibility menu, dialog, popup

UserName

CAS RN 2374-14-3

CH3SiFFFOSiCH3FFFOSiH3CFFFO

CAS RN 2374-14-3

IUPAC Name: 2,4,6-trimethyl-2,4,6-tris(3,3,3-trifluoropropyl)-1,3,5,2,4,6-trioxatrisilinane
Synonyms: 2,4,6-trimethyl-2,4,6-tris(3,3,3-trifluoropropyl)-1,3,5,2,4,6-trioxatrisilinane
Molecular Weight (g/mol): 468.54
Molecular Formula: C12H21F9O3Si3
InChi Key: URZHQOCYXDNFGN-UHFFFAOYSA-N
SMILES: C[Si]1(CCC(F)(F)F)O[Si](C)(CCC(F)(F)F)O[Si](C)(CCC(F)(F)F)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results