CAS RN 2373-89-9
IUPAC Name:
(2E)-1,3-bis(4-methoxyphenyl)prop-2-en-1-one
Synonyms:
(2E)-1,3-bis(4-methoxyphenyl)prop-2-en-1-one
Molecular Weight (g/mol):
268.31
Molecular Formula:
C17H16O3
InChi Key:
HDXVSZWKIHQDES-LFYBBSHMSA-N
SMILES:
COC1=CC=C(\C=C\C(=O)C2=CC=C(OC)C=C2)C=C1
1
–
1
of
1
results
1
4,4'-Dimethoxychalcone, 99%
CAS: 2373-89-9 Molecular Formula: C17H16O3 Molecular Weight (g/mol): 268.31 MDL Number: MFCD00025815 InChI Key: HDXVSZWKIHQDES-LFYBBSHMSA-N Synonym: 4,4'-dimethoxychalcone,e-1,3-bis 4-methoxyphenyl prop-2-en-1-one,2e-1,3-bis 4-methoxyphenyl prop-2-en-1-one,1,3-bis 4-methoxyphenyl prop-2-en-1-one,4,4/'-dimethoxychalcone,2-propen-1-one, 1,3-bis 4-methoxyphenyl,e-4,4'-dimethoxychalcone,1,3-bis 4-methoxyphenyl-2-propen-1-one,e-1,3-bis 4-methoxyphenyl-2-propen-1-one,2e-1,3-bis 4-methoxyphenyl-2-propen-1-one PubChem CID: 5377817 IUPAC Name: (E)-1,3-bis(4-methoxyphenyl)prop-2-en-1-one SMILES: COC1=CC=C(\C=C\C(=O)C2=CC=C(OC)C=C2)C=C1