accessibility menu, dialog, popup

UserName

NH2FNOO

CAS RN 2369-13-3

IUPAC Name: 3-fluoro-4-nitroaniline
Synonyms: 3-fluoro-4-nitroaniline
Molecular Weight (g/mol): 156.12
Molecular Formula: C6H5FN2O2
InChi Key: KKQPNAPYVIIXFB-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(F)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results