accessibility menu, dialog, popup

UserName

CAS RN 23674-20-6

Br

CAS RN 23674-20-6

IUPAC Name: 9-bromo-10-phenylanthracene
Synonyms: 9-bromo-10-phenylanthracene
Molecular Weight (g/mol): 333.23
Molecular Formula: C20H13Br
InChi Key: WHGGVVHVBFMGSG-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results