CAS RN 23642-66-2
CAS RN 23642-66-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Benfluorex Hydrochloride, TRC
CAS: 23642-66-2 Molecular Formula: C19 H20 F3 N O2 . Cl H Synonym: Ethanol, 2-[[1-methyl-2-[3-(trifluoromethyl)phenyl]ethyl]amino]-, 1-benzoate, hydrochloride (1:1)∣Ethanol, 2-[[1-methyl-2-[3-(trifluoromethyl)phenyl]ethyl]amino]-, benzoate (ester), hydrochloride (9CI)∣Ethanol, 2-[[α-methyl-m-(trifluoromethyl)phenethyl]amino]-, benzoate (ester), hydrochloride (8CI)∣(±)-N-(2-Benzoyloxyethyl)norfenfluramine hydrochloride∣Benfluorex hydrochloride∣JP 992∣Mediator∣Mediaxal∣S 992