CAS RN 23628-31-1
CAS RN 23628-31-1
IUPAC Name:
6-aminopyridine-2-carboxylic acid
Synonyms:
6-aminopyridine-2-carboxylic acid
Molecular Weight (g/mol):
138.13
Molecular Formula:
C6H6N2O2
InChi Key:
NMCKJFCJIHCHIS-UHFFFAOYSA-N
SMILES:
NC1=CC=CC(=N1)C(O)=O
1
–
1
of
1
results
1
6-Aminopyridine-2-carboxylic acid, 95%
CAS: 23628-31-1 Molecular Formula: C6H6N2O2 Molecular Weight (g/mol): 138.13 MDL Number: MFCD00233711 InChI Key: NMCKJFCJIHCHIS-UHFFFAOYSA-N Synonym: 6-aminopicolinic acid,2-aminopyridine-6-carboxylic acid,6-amino-2-pyridinecarboxylic acid,6-amino-2-picolinic acid,6-amino-pyridine-2-carboxylic acid,2-pyridinecarboxylic acid, 6-amino,2-amino-6-pyridinecarboxylic acid,picolinic acid, 6-amino,pubchem7686,acmc-209g4h PubChem CID: 351652 IUPAC Name: 6-aminopyridine-2-carboxylic acid SMILES: NC1=CC=CC(=N1)C(O)=O