accessibility menu, dialog, popup

UserName

H3COBrF

CAS RN 2357-52-0

IUPAC Name: 4-bromo-2-fluoro-1-methoxybenzene
Synonyms: 4-bromo-2-fluoro-1-methoxybenzene
Molecular Weight (g/mol): 205.03
Molecular Formula: C7H6BrFO
InChi Key: DWNXGZBXFDNKOR-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results