accessibility menu, dialog, popup

UserName

CAS RN 2346-00-1

IUPAC Name: 2-methyl-4,5-dihydro-1,3-thiazole
Synonyms: 2-methyl-4,5-dihydro-1,3-thiazole
Molecular Weight (g/mol): 101.17
Molecular Formula: C4H7NS
InChi Key: JUIQOABNSLTJSW-UHFFFAOYSA-N
SMILES: CC1=NCCS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results