CAS RN 23312-56-3
CAS RN 23312-56-3
IUPAC Name:
(1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
Synonyms:
spectinomycin
Molecular Weight (g/mol):
430.43
Molecular Formula:
C14H26N2O11S
InChi Key:
XGBFWQUQYQIFLB-MTTMTQIXSA-N
SMILES:
OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
1
–
1
of
1
results
1
Spectinomycin Sulfate, ≥98%, MP Biomedicals™
CAS: 23312-56-3 Molecular Formula: C14H26N2O11S Molecular Weight (g/mol): 430.43 MDL Number: MFCD00270185 InChI Key: XGBFWQUQYQIFLB-MTTMTQIXSA-N Synonym: U 18409E, antibiotic ChEBI: CHEBI:9216 IUPAC Name: (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid SMILES: OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O