CAS RN 2319722-40-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ GYKI 52466 dihydrochloride
Tocris Bioscience™ GYKI 52466 dihydrochloride
CAS: 2319722-40-0 Molecular Formula: C17H15N3O2