accessibility menu, dialog, popup

UserName

CAS RN 23190-16-1

HONH2

CAS RN 23190-16-1

IUPAC Name: 2-amino-1,2-diphenylethan-1-ol
Synonyms: ethanol, 2-amino-1,2-diphenyl-
Molecular Weight (g/mol): 213.28
Molecular Formula: C14H15NO
InChi Key: GEJJWYZZKKKSEV-UHFFFAOYNA-N
SMILES: NC(C(O)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results