accessibility menu, dialog, popup

UserName

INCO

CAS RN 23138-56-9

IUPAC Name: 1-iodo-3-isocyanatobenzene
Synonyms: 1-iodo-3-isocyanatobenzene
Molecular Weight (g/mol): 245.02
Molecular Formula: C7H4INO
InChi Key: LZSQANBVKPXBLO-UHFFFAOYSA-N
SMILES: IC1=CC=CC(=C1)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results