CAS RN 23103-35-7
CAS RN 23103-35-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
myo-Inositol Trispyrophosphate Hexasodium Salt, TRC
CAS: 23103-35-7 Molecular Formula: C6 H6 O21 P6 . 6[Na] Molecular Weight (g/mol): 737.88 Synonym: ITTP Hexasodium Salt,myo-Inositol Cyclic 1,2:3,4:5,6-Tris(dihydrogen pyrophosphate)) Hexasodium Salt,myo-Inositol Cyclic 1,2:3,4:5,6-Tris(P,P'-dihydrogen diphosphate) Hexasodium Salt SMILES: O=P1(O[C@]2([H])[C@@]3([H])[C@@](OP(OP(O3)(O[Na])=O)(O[Na])=O)([H])[C@]4([H])[C@@](OP(OP(O4)(O[Na])=O)(O[Na])=O)([H])[C@@]2([H])OP(O1)(O[Na])=O)O[Na]