accessibility menu, dialog, popup

UserName

CAS RN 23062-17-1

IUPAC Name: 2-methyl-1,3-oxazole-4-carboxylic acid
Synonyms: 2-methyl-1,3-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 127.1
Molecular Formula: C5H5NO3
InChi Key: IARMCEYEYXXEOS-UHFFFAOYSA-N
SMILES: CC1=NC(=CO1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results