accessibility menu, dialog, popup

UserName

CAS RN 23056-39-5

CH3NClNOO

CAS RN 23056-39-5

IUPAC Name: 2-chloro-4-methyl-3-nitropyridine
Synonyms: 2-chloro-4-methyl-3-nitropyridine
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: JHARVUVBTAAPLA-UHFFFAOYSA-N
SMILES: CC1=CC=NC(Cl)=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)