accessibility menu, dialog, popup

UserName

CAS RN 2304-81-6

H3CONHOHCH3OH(E/Z)

CAS RN 2304-81-6

IUPAC Name: N-(1,3-dihydroxyoctadec-4-en-2-yl)octadecanamide
Synonyms: N-(1,3-dihydroxyoctadec-4-en-2-yl)octadecanamide
Molecular Weight (g/mol): 565.97
Molecular Formula: C36H71NO3
InChi Key: VODZWWMEJITOND-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)NC(CO)C(O)C=CCCCCCCCCCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results