accessibility menu, dialog, popup

UserName

OIF

CAS RN 227609-88-3

IUPAC Name: 4-fluoro-3-iodobenzaldehyde
Synonyms: 4-fluoro-3-iodobenzaldehyde
Molecular Weight (g/mol): 250.01
Molecular Formula: C7H4FIO
InChi Key: DGUYWJGDFCRUHC-UHFFFAOYSA-N
SMILES: FC1=C(I)C=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results