accessibility menu, dialog, popup

UserName

CAS RN 2273-51-0

OSn

CAS RN 2273-51-0

IUPAC Name: diphenylstannanone
Synonyms: diphenylstannanone
Molecular Weight (g/mol): 288.92
Molecular Formula: C12H10OSn
InChi Key: VPPWQRIBARKZNY-UHFFFAOYSA-N
SMILES: O=[Sn](C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results