CAS RN 2268-05-5
CAS RN 2268-05-5
IUPAC Name:
1,3-dichloro-2-fluorobenzene
Synonyms:
1,3-dichloro-2-fluorobenzene
Molecular Weight (g/mol):
164.99
Molecular Formula:
C6H3Cl2F
InChi Key:
JORVCRLRRRRLFI-UHFFFAOYSA-N
SMILES:
FC1=C(Cl)C=CC=C1Cl
1
–
1
of
1
results
1
1,3-Dichloro-2-fluorobenzene, 98%, Thermo Scientific™
CAS: 2268-05-5 MDL Number: MFCD00010307