CAS RN 22610-99-7
CAS RN 22610-99-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
cis-Diethyl Stilbestrol, TRC
CAS: 22610-99-7 Molecular Formula: C18 H20 O2 Molecular Weight (g/mol): 268.35 Synonym: Phenol, 4,4'-[(1Z)-1,2-diethyl-1,2-ethenediyl]bis-,4,4'-Stilbenediol, α,α'-diethyl-, (Z)- (8CI),Phenol, 4,4'-(1,2-diethyl-1,2-ethenediyl)bis-, (Z)-,4,4'-[(1Z)-1,2-Diethyl-1,2-ethenediyl]bis[phenol],(Z)-3,4-Bis(4-hydroxyphenyl)-3-hexene,(Z)-DES,cis-Diethylstilbestrol,(Z)-Diethylstilbestrol IUPAC Name: 4-[(Z)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol SMILES: CC\C(=C(/CC)\c1ccc(O)cc1)\c2ccc(O)cc2