accessibility menu, dialog, popup

UserName

CAS RN 22581-63-1

BrBr2H2H2H2H

CAS RN 22581-63-1

IUPAC Name: dibromo(²H₄)ethane
Synonyms: dibromo(²H₄)ethane
Molecular Weight (g/mol): 191.89
Molecular Formula: C2H4Br2
InChi Key: PAAZPARNPHGIKF-LNLMKGTHSA-N
SMILES: [2H]C([2H])(Br)C([2H])([2H])Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results