CAS RN 2246614-97-9
CAS RN 2246614-97-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalonitrile (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalonitrile (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.