accessibility menu, dialog, popup

UserName

CAS RN 2240-88-2

OHFFF

CAS RN 2240-88-2

IUPAC Name: 3,3,3-trifluoropropan-1-ol
Synonyms: 3,3,3-trifluoro-1-propanol
Molecular Weight (g/mol): 114.07
Molecular Formula: C3H5F3O
InChi Key: HDBGBTNNPRCVND-UHFFFAOYSA-N
SMILES: OCCC(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results