accessibility menu, dialog, popup

UserName

NClNH2

CAS RN 22353-34-0

IUPAC Name: 5-chloropyridin-3-amine
Synonyms: 5-chloropyridin-3-amine
Molecular Weight (g/mol): 128.56
Molecular Formula: C5H5ClN2
InChi Key: ZHMASVAJFJFFLS-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CN=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)