accessibility menu, dialog, popup

UserName

CAS RN 22282-72-0

HOOONH

CAS RN 22282-72-0

IUPAC Name: 2-oxo-1,2-dihydropyridine-4-carboxylic acid
Synonyms: 2-oxo-1H-pyridine-4-carboxylic acid
Molecular Weight (g/mol): 139.11
Molecular Formula: C6H5NO3
InChi Key: BXHCJLRTXPHUGH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results