accessibility menu, dialog, popup

UserName

CAS RN 2226-96-2

CH3H3COHH3CH3CNO

CAS RN 2226-96-2

IUPAC Name: (4-hydroxy-2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Synonyms: 4-Hydroxy-TEMPO
Molecular Weight (g/mol): 172.25
Molecular Formula: C9H18NO2
InChi Key: UZFMOKQJFYMBGY-UHFFFAOYSA-N
SMILES: CC1(C)CC(O)CC(C)(C)N1[O]
Quantity 
  • (4)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)