accessibility menu, dialog, popup

UserName

CAS RN 22112-78-3

CH3ONHOH3CNOH3CNHNOCH3

CAS RN 22112-78-3

IUPAC Name: 2,7,12,17-tetrakis(4-methoxyphenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Synonyms: 2,7,12,17-tetrakis(4-methoxyphenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Molecular Weight (g/mol): 734.86
Molecular Formula: C48H38N4O4
InChi Key: SKXWDGDNWBYACJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results