accessibility menu, dialog, popup

UserName

CAS RN 221006-70-8

H3COBOHHOOH3CN

CAS RN 221006-70-8

IUPAC Name: (2,6-dimethoxypyridin-3-yl)boronic acid
Synonyms: 2,6-dimethoxypyridin-3-ylboronic acid
Molecular Weight (g/mol): 182.97
Molecular Formula: C7H10BNO4
InChi Key: ADGHSWFUZUADDH-UHFFFAOYSA-N
SMILES: COC1=CC=C(B(O)O)C(OC)=N1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)