CAS RN 2210-79-9
IUPAC Name:
2-[(2-methylphenoxy)methyl]oxirane
Synonyms:
cresyl glycidyl ether
Molecular Weight (g/mol):
164.2
Molecular Formula:
C10H12O2
InChi Key:
KFUSXMDYOPXKKT-UHFFFAOYNA-N
SMILES:
CC1=CC=CC=C1OCC1CO1
1
–
1
of
1
results
1
Glycidyl 2-methylphenyl ether, tech. 85%
CAS: 2210-79-9 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.204 MDL Number: MFCD00037847 InChI Key: KFUSXMDYOPXKKT-UHFFFAOYSA-N Synonym: 2-2-methylphenoxy methyl oxirane,o-cresyl glycidyl ether,glycidyl 2-methylphenyl ether,o-cresol glycidyl ether,glycidyl o-tolyl ether,araldite dy 023,o-kresol-glycidaether,2-o-tolyloxy methyl oxirane,1,2-epoxy-3-2-methylphenoxy propane,1-2-methylphenoxy-2,3-epoxypropane PubChem CID: 16640 IUPAC Name: 2-[(2-methylphenoxy)methyl]oxirane SMILES: CC1=CC=CC=C1OCC2CO2