accessibility menu, dialog, popup

UserName

CAS RN 219967-69-8

HN(S)OOOOHOH3CCH3H3C

CAS RN 219967-69-8

IUPAC Name: (3S)-4-[4-(tert-butoxy)phenyl]-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: (3S)-4-[4-(tert-butoxy)phenyl]-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 473.57
Molecular Formula: C29H31NO5
InChi Key: PAFSELQEYJTCCG-FQEVSTJZSA-N
SMILES: CC(C)(C)OC1=CC=C(C[C@@H](CC(O)=O)NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results