accessibility menu, dialog, popup

UserName

CAS RN 21901-40-6

CH3NH2NHNOO

CAS RN 21901-40-6

IUPAC Name: 2-amino-4-methyl-5-nitropyridin-1-ium
Synonyms: 2-amino-4-methyl-5-nitropyridin-1-ium
Molecular Weight (g/mol): 154.15
Molecular Formula: C6H8N3O2
InChi Key: GRBBNZYMXKTQAI-UHFFFAOYSA-O
SMILES: CC1=CC(N)=[NH+]C=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)