accessibility menu, dialog, popup

UserName

CAS RN 21643-38-9

H3COHO

CAS RN 21643-38-9

IUPAC Name: 4-hexylbenzoic acid
Synonyms: benzoic acid, 4-hexyl-
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: CPEPWESLFZVUEP-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)