CAS RN 21642-98-8
CAS RN 21642-98-8
IUPAC Name:
4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile
Synonyms:
4-methoxy-2-oxo-1H-pyridine-3-carbonitrile
Molecular Weight (g/mol):
150.14
Molecular Formula:
C7H6N2O2
InChi Key:
MWGIDWPSRDMIQN-UHFFFAOYSA-N
SMILES:
COC1=C(C#N)C(=O)NC=C1
1
–
2
of
2
results
1
3-Cyano-4-methoxy-2-pyridone, 95%, Thermo Scientific Chemicals
CAS: 21642-98-8 Molecular Formula: C7H6N2O2 Molecular Weight (g/mol): 150.14 MDL Number: MFCD00975442 InChI Key: MWGIDWPSRDMIQN-UHFFFAOYSA-N Synonym: 4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile,4-methoxy-2-oxo-1,2-dihydro-pyridine-3-carbonitrile,3-cyano-2-hydroxy-4-methoxypyridine,4-methoxy-2-oxo-1,2-dihydro-3-pyridinecarbonitrile,2-hydroxy-4-methoxynicotinonitrile,1,2-dihydro-4-methoxy-2-oxo-3-pyridinecarbonitrile,3-pyridinecarbonitrile, 1,2-dihydro-4-methoxy-2-oxo,3-cyano-4-methoxy-2 1h-pyridinone,2-hydroxy-4-methoxy-nicotinonitrile,3-cyano-4-methoxy-2-1h-pyridinone PubChem CID: 2786702 SMILES: COC1=C(C#N)C(=O)NC=C1