accessibility menu, dialog, popup

UserName

CAS RN 2156-04-9

IUPAC Name: (4-ethenylphenyl)boronic acid
Synonyms: 4-ethenylphenylboronic acid
Molecular Weight (g/mol): 147.97
Molecular Formula: C8H9BO2
InChi Key: QWMJEUJXWVZSAG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results